missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fast Blue BB salt

Product Code. 11372217
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11372217

Brand: Thermo Scientific Chemicals L09704.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5486-84-0 | C34H36Cl4N6O6Zn | 831.878 g/mol

Fast Blue BB salt is used as a general substrate for β-glycosidase. Two moles of 6-bromo-2-napthol-β-d-glucopyranoside are cleaved by β-glucosidases into 2 moles of d-glucose and 2 moles of 6-bromo-2-napthol. The 2 moles of 6-bromo-2-napthol formed are coupled with 1 mole of Fast Blue BB salt [4-benzoylamino-2,5-diethoxybenzenediazonium chloride hemi (zinc chloride) salt], resulting in 2 moles of the corresponding dye of 6-bromo-2-naphthol (reddish color). It is a commonly used stain to observe myelin under light microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fast Blue BB salt is used as a general substrate for β-glycosidase. Two moles of 6-bromo-2-napthol-β-d-glucopyranoside are cleaved by β-glucosidases into 2 moles of d-glucose and 2 moles of 6-bromo-2-napthol. The 2 moles of 6-bromo-2-napthol formed are coupled with 1 mole of Fast Blue BB salt [4-benzoylamino-2,5-diethoxybenzenediazonium chloride hemi (zinc chloride) salt], resulting in 2 moles of the corresponding dye of 6-bromo-2-naphthol (reddish color). It is a commonly used stain to observe myelin under light microscopy.

Solubility
Soluble in water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5486-84-0
Molecular Formula C34H36Cl4N6O6Zn
Molecular Weight (g/mol) 831.878
MDL Number MFCD00074765
InChI Key CMFRFQODFZBKTI-UHFFFAOYSA-L
Synonym 4-Benzamido-2,5-diethoxybenzene diazonium chloride hemi zinc salt; C.I. 37175
PubChem CID 131675364
IUPAC Name zinc;N-(4-diazonio-2,5-diethoxyphenyl)benzenecarboximidate;dichloride;dihydrochloride
SMILES CCOC1=CC(=C(C=C1N=C(C2=CC=CC=C2)[O-])OCC)[N+]#N.CCOC1=CC(=C(C=C1N=C(C2=CC=CC=C2)[O-])OCC)[N+]#N.Cl.Cl.[Cl-].[Cl-].[Zn+2]

Specifications

Melting Point ∼157°C (decomposition)
Beilstein 1845288
Solubility Information Soluble in water.
Formula Weight 831.89
Odor Odorless
Sensitivity Light sensitive
Quantity 5 g
Chemical Name or Material Fast Blue BB salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.