missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fast Blue BB salt

Product Code. 11362217
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362217 1 g 1g
11372217 5 g 5g
11382217 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11362217 Supplier Thermo Scientific Chemicals Supplier No. L09704.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 5486-84-0 | C34H36Cl4N6O6Zn | 831.878 g/mol

Fast Blue BB salt is used as a general substrate for β-glycosidase. Two moles of 6-bromo-2-napthol-β-d-glucopyranoside are cleaved by β-glucosidases into 2 moles of d-glucose and 2 moles of 6-bromo-2-napthol. The 2 moles of 6-bromo-2-napthol formed are coupled with 1 mole of Fast Blue BB salt [4-benzoylamino-2,5-diethoxybenzenediazonium chloride hemi (zinc chloride) salt], resulting in 2 moles of the corresponding dye of 6-bromo-2-naphthol (reddish color). It is a commonly used stain to observe myelin under light microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fast Blue BB salt is used as a general substrate for β-glycosidase. Two moles of 6-bromo-2-napthol-β-d-glucopyranoside are cleaved by β-glucosidases into 2 moles of d-glucose and 2 moles of 6-bromo-2-napthol. The 2 moles of 6-bromo-2-napthol formed are coupled with 1 mole of Fast Blue BB salt [4-benzoylamino-2,5-diethoxybenzenediazonium chloride hemi (zinc chloride) salt], resulting in 2 moles of the corresponding dye of 6-bromo-2-naphthol (reddish color). It is a commonly used stain to observe myelin under light microscopy.

Solubility
Soluble in water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5486-84-0
Molecular Formula C34H36Cl4N6O6Zn
Molecular Weight (g/mol) 831.878
MDL Number MFCD00074765
InChI Key CMFRFQODFZBKTI-UHFFFAOYSA-L
Synonym 4-Benzamido-2,5-diethoxybenzene diazonium chloride hemi zinc salt; C.I. 37175
PubChem CID 131675364
IUPAC Name zinc;N-(4-diazonio-2,5-diethoxyphenyl)benzenecarboximidate;dichloride;dihydrochloride
SMILES CCOC1=CC(=C(C=C1N=C(C2=CC=CC=C2)[O-])OCC)[N+]#N.CCOC1=CC(=C(C=C1N=C(C2=CC=CC=C2)[O-])OCC)[N+]#N.Cl.Cl.[Cl-].[Cl-].[Zn+2]

Specifications

Melting Point ∼157°C (decomposition)
Beilstein 1845288
Solubility Information Soluble in water.
Formula Weight 831.89
Odor Odorless
Sensitivity Light sensitive
Quantity 1 g
Chemical Name or Material Fast Blue BB salt
compliance-icons
Product Identifier
  • Fast Blue BB salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Fast Blue BB salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.