missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fast Blue BB base

Product Code. 15474829
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15474829 25 g 25g
15484829 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15474829 Supplier Thermo Scientific Chemicals Supplier No. J62456.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 120-00-3 | C17H20N2O3 | 300.358 g/mol

C.I. 37175

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C.I. 37175
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-00-3
Molecular Formula C17H20N2O3
Molecular Weight (g/mol) 300.358
MDL Number MFCD00009091
InChI Key CNXZLZNEIYFZGU-UHFFFAOYSA-N
Synonym C.I. 37175; 4'-Amino-2',5'-diethoxybenzanilide
PubChem CID 67108
IUPAC Name N-(4-amino-2,5-diethoxyphenyl)benzamide
SMILES CCOC1=CC(=C(C=C1N)OCC)NC(=O)C2=CC=CC=C2

Specifications

Melting Point 95°C to 105°C
Beilstein 3416889
Formula Weight 300.35
Color Brown to Gray
Quantity 25 g
Physical Form Powder
Chemical Name or Material Fast Blue BB base

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.