missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FASN Monoclonal Antibody (C.60.10)

Rabbit Monoclonal Antibody
Brand: Invitrogen™ MA514887
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
Fatty acid synthase is a multifunctional protein. Its main function is to catalyze the synthesis of palmitate from acetyl-CoA and malonyl-CoA, in the presence of NADPH, into long-chain saturated fatty acids. In some cancer cell lines, this protein has been found to be fused with estrogen receptor-alpha (ER-alpha), in which the N-terminus of FAS is fused in-frame with the C-terminus of ER-alpha.
Specifications
| FASN | |
| Monoclonal | |
| 5 μg/mL | |
| 0.01M HEPES with 0.15M NaCl, 50% glycerol, 100 μg/mL BSA and <0.02% sodium azide; pH 7.5 | |
| P12785, P19096, P49327 | |
| FASN | |
| Synthetic peptide around Gly46 corresponding to the sequence of human fatty acid synthase. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| C.60.10 | |
| Unconjugated | |
| FASN | |
| [Acyl-carrier-protein] S-acetyltransferase; [Acyl-carrier-protein] S-malonyltransferase; 3-hydroxyacyl-[acyl-carrier-protein] dehydratase; 3-oxoacyl-[acyl-carrier-protein] reductase; 3-oxoacyl-[acyl-carrier-protein] synthase; A630082H08Rik; Enoyl-[acyl-carrier-protein] reductase; FAS; FASN; Fatty acid synthase; fatty acid synthase (EC 3.2.1.23); MGC14367; MGC15706; OA-519; Oleoyl-[acyl-carrier-protein] hydrolase; SDR27X1; short chain dehydrogenase/reductase family 27X, member 1; thioesterase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14104, 2194, 50671 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction