missing translation for 'onlineSavingsMsg'
Learn More

Fasiglifam, MedChemExpress

Product Code. 16466707
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16466707

Brand: Medchem Express HY10480

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fasiglifam (TAK-875) is a potent, selective and orally bioavailable GPR40 agonist with EC50 of 72 nM.

Fasiglifam (TAK-875) is a potent, selective and orally bioavailable GPR40 agonist with EC50 of 72 nM.

Specifications

Chemical Name or Material Fasiglifam
CAS 1000413-72-8
Color Red
Purity Grade Notes Research
Molecular Formula C29H32O7S
Quantity 5 mg
Synonym TAK-875
Solubility Information DMSO : ≥ 128 mg/mL (243.98 mM)
SMILES CC1=C(C(C)=CC(OCCCS(C)(=O)=O)=C1)C2=CC(COC3=CC=C4[C@@H](COC4=C3)CC(O)=O)=CC=C2
Molecular Weight (g/mol) 524.63
Formula Weight 524.63
Percent Purity 95.05%
Grade Research
Physical Form Liquid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.