missing translation for 'onlineSavingsMsg'
Learn More

Fasiglifam, MedChemExpress

Product Code. 16490347
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490347 50 mg 50mg
16484996 100 mg 100mg
16443998 10 mg 10mg
16466707 5 mg 5mg
16420116 2 mg 2mg
16480986 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16490347 Supplier Medchem Express Supplier No. HY10480
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Fasiglifam (TAK-875) is a potent, selective and orally bioavailable GPR40 agonist with EC50 of 72 nM.

Fasiglifam (TAK-875) is a potent, selective and orally bioavailable GPR40 agonist with EC50 of 72 nM.

Specifications

Chemical Name or Material Fasiglifam
CAS 1000413-72-8
Color Red
Purity Grade Notes Research
Molecular Formula C29H32O7S
Quantity 50 mg
Synonym TAK-875
Solubility Information DMSO : ≥ 128 mg/mL (243.98 mM)
SMILES CC1=C(C(C)=CC(OCCCS(C)(=O)=O)=C1)C2=CC(COC3=CC=C4[C@@H](COC4=C3)CC(O)=O)=CC=C2
Molecular Weight (g/mol) 524.63
Formula Weight 524.63
Percent Purity 95.05%
Grade Research
Physical Form Liquid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.