missing translation for 'onlineSavingsMsg'
Learn More

Fasentin, TRC

Product Code. 30390594
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390594 100 mg
30396934 10 mg
30402652 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390594 Supplier Toronto Research Chemicals Supplier No. TRCF150020100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 392721-37-8
Molecular Formula C11H9ClF3NO2
Molecular Weight (g/mol) 279.64
Synonym N-(4-Chloro-3-(trifluoromethyl)phenyl)-3-oxobutanamide
IUPAC Name N-[4-chloro-3-(trifluoromethyl)phenyl]-3-oxobutanamide
SMILES CC(CC(NC1=CC(C(F)(F)F)=C(Cl)C=C1)=O)=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H9ClF3NO2/c1-6(17)4-10(18)16-7-2-3-9(12)8(5-7)11(13,14)15/h2-3,5H,4H2,1H3,(H,16,18)
Formula Weight 279.0274
Chemical Name or Material Fasentin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.