missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fascin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14384-1-AP-150UL
This item is not returnable.
View return policy
Description
Human fascin is a highly conserved actin-bundling protein. Fascin, encoded by the human homolog for sn (hsn) gene, has been localized to microspikes and stress fibers of cultured cells where it is thought to be involved in the formation of microfilament bundles. It is expressed predomitly in dendritic cells. Lymphoid cells, myeloid cells and plasma cells are negative. However. Reed Sternberg cells in Hodgkin′s lymphoma are positive for fascin staining. Epstein-Barr virus may induce expression of fascin in B cells.Specifications
| Fascin | |
| Polyclonal | |
| Unconjugated | |
| FSCN1 | |
| 55 kDa actin bundling protein, FAN1, Fascin, Fascin 1,Fascin, FSCN1, HSN, p55, Singed like protein, SNL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14086, 6624 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q16658, Q61553 | |
| FSCN1 | |
| Fascin Fusion Protein Ag5816 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction