missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fascin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116442
This item is not returnable.
View return policy
Description
Positive controls: SH-SY5Y, rat brain tissue, human tonsil tissue, human kidney tissue, mouse testis tissue.
Fascin organizes filamentous actin into bundles with a minimum of 4. 1:1 actin/fascin ratio. It is probably involved in the assembly of actin filament bundles present in microspikes, membrane ruffles, and stress fibers.
Specifications
| Fascin | |
| Polyclonal | |
| Unconjugated | |
| FSCN1 | |
| 55 kDa actin-bundling protein; 55kDa actin-bundling protein; AI663989; Fan1; fascin; fascin 1; fascin actin-bundling protein 1; Fascin homolog 1; fascin homolog 1, actin bundling protein; fascin homolog 1, actin bundling protein (Strongylocentrotus purpuratus); fascin homolog 1, actin-bundling protein; Fascin1; fascin-1; Fscn1; fscn1 {ECO:0000250; HSN; p55; Singed; Singed (Drosophila) like (sea urchin fascin homolog like); singed-like (fascin homolog, sea urchin); singed-like protein; SNL; UniProtKB:Q16658} | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14086, 6624, 683788 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| P85845, Q16658, Q61553 | |
| FSCN1 | |
| Recombinant protein within Human Fascin aa 1-140. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction