missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fascin Monoclonal Antibody (FCN01 (55K-2)), Invitrogen™
Description
MA5-11483 targets Fascin in IHC (P) applications and shows reactivity with Human and Rat samples. The MA5-11483 immunogen is fascin purified from HeLa cells.
Human fascin is a highly conserved actin-bundling protein. Fascin, encoded by the human homolog for sn (hsn) gene, has been localized to microspikes and stress fibers of cultured cells where it is thought to be involved in the formation of microfilament bundles. It is expressed predomitly in dendritic cells. Lymphoid cells, myeloid cells and plasma cells are negative. However. Reed Sternberg cells in Hodgkin′s lymphoma are positive for fascin staining. Epstein-Barr virus may induce expression of fascin in B cells.
Specifications
Specifications
| Antigen | Fascin |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | FCN01 (55K-2) |
| Conjugate | Unconjugated |
| Formulation | tissue culture supernatant with 0.09% sodium azide |
| Gene | FSCN1 |
| Gene Accession No. | Q16658, Q61553, P85845 |
| Gene Alias | 55 kDa actin-bundling protein, Singed, drosophila, homolog-like, actin bundling protein, fascin, singed-like (fascin homolog, sea urchin), singed-like protein |
| Gene Symbols | FSCN1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?