missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Fascin Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Fascin Ab-1, Mouse Monoclonal Antibody.

£167.00 - £605.00

Specifications

Antigen Fascin Ab-1
Clone FCN01; same as 55K-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Quantity Form Price Quantity & Availability  
Product Code Brand Quantity Form Price Quantity & Availability  
12658996
View Documents
Epredia™
MS-1112-S0
100 μL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12668996
View Documents
Epredia™
MS-1112-S1
500 μL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12648996
View Documents
Epredia™
MS-1112-S
1 mL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12628996
View Documents
Epredia™
MS-1112-R7
7 mL Prediluted This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12638996
View Documents
Epredia™
MS-1112-RQ
12 mL Ready-to-Use This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Human fascin is a highly conserved actin-bundling protein. Fascin, encoded by the human homolog for sn (hsn) gene, has been localized to microspikes and stress fibers of cultured cells where it is thought to be involved in the formation of microfilament bundles. It is expressed predominantly in dendritic cells. Lymphoid cells, myeloid cells and plasma cells are negative. However, Reed Sternberg cells in Hodgkin's lymphoma are positive for fascin staining. Epstein-Barr virus may induce expression of fascin in B cells.

Host Species: Mouse

Clone: FCN01; same as 55K-2

Isotype: IgG1

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Fascin purified from HeLa cells

Molecular Weight: 55 kDa

Positive Control: Hodgkin's lymphoma or spleen

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

Fascin Ab-1
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Fascin purified from HeLa cells
FCN01; same as 55K-2
Monoclonal
Mouse
Neuroscience
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.