missing translation for 'onlineSavingsMsg'
Learn More

Fas/TNFRSF6/CD95 Mouse anti-Human, Clone: B-G27, Novus Biologicals™

Product Code. 18336235 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
200 μg
500 μg
Unit Size:
200µL
500µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18336235 200 μg 200µL
18316756 500 μg 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18336235 Supplier Novus Biologicals Supplier No. NBP314592200UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Fas/TNFRSF6/CD95 Monoclonal antibody specifically detects Fas/TNFRSF6/CD95 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fas/TNFRSF6/CD95
Applications Flow Cytometry, Immunohistochemistry, Functional Assay
Classification Monoclonal
Clone B-G27
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, Immunohistochemistry, Functional
Formulation PBS. Sterile-filtered through 0.22 um.
Gene Alias Apo-1 antigen, apoptosis antigen 1, Apoptosis-mediating surface antigen FAS, APT1FASTM, CD95 antigen, CD95ALPS1A, Fas (TNF receptor superfamily, member 6), Fas AMA, Fas antigen, FAS1, FASLG receptor, TNFRSF6member 6, tumor necrosis factor receptor superfamily member 6
Host Species Mouse
Immunogen Recombinant Fas/TNFRSF6/CD95 antigen
Purification Method Affinity purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Death Receptor Signaling Pathway, Diabetes Research, Hematopoietic Stem Cell Markers, Immunology, Inhibitors Activators and Regulators, Innate Immunity, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 355
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.