missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ FARSLA Antibody, Novus Biologicals™

Product Code. 18343978 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18343978 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18343978 Supplier Novus Biologicals™ Supplier No. H00002193B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

FARSLA Polyclonal antibody specifically detects FARSLA in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen FARSLA
Applications Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_004452.1
Gene Alias CML33pheRS, EC 6.1.1.20, FARS, FARSL, FARSLA, FRSA, PheHA, phenylalanine tRNA ligase 1, alpha, cytoplasmic, Phenylalanine--tRNA ligase alpha chain, phenylalanine-tRNA synthetase alpha-subunit, phenylalanine-tRNA synthetase-like, phenylalanine-tRNA synthetase-like, alpha subunit, phenylalanyl-tRNA synthetase alpha chain, phenylalanyl-tRNA synthetase, alpha subunit, phenylalanyl-tRNA synthetase-like, alpha subunit, PheRS
Host Species Mouse
Immunogen FARSA (NP_004452.1, 1 a.a. - 508 a.a.) full-length human protein. MADGQVAELLLRRLEASDGGLDSAELAAELGMEHQAVVGAVKSLQALGEVIEAELRSTKHWELTAEGEEIAREGSHEARVFRSIPPEGLAQSELMRLPSGKVGFSKAMSNKWIRVDKSAADGPRVFRVVDSMEDEVQRRLQLVRGGQAEKLGEKERSELRKRKLLAEVTLKTYWVSKGSAFSTSISKQETELSPEMISSGSWRDRPFKPYNFLAHGVLPDSGHLHPLLKVRSQFRQIFLEMGFTEMPTDNFIESSFWNFDALFQPQQHPARDQHDTFFLRDPAEALQLPMDYVQRVKRTHSQGGYGSQGYKYNWKLDEARKNLLRTHTTSASARALYRLAQKKPFTPVKYFSIDRVFRNETLDATHLAEFHQIEGVVADHGLTLGHLMGVLREFFTKLGITQLRFKPAYNPYTEPSMEVFSYHQGLKKWVEVGNSGVFRPEMLLPMGLPENVSVIAWGLSLERPTMIKYGINNIRELVGHKVNLQMVYDSPLCRLDAEPRPPPTQEAA
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2193
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.