missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FARSB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | FARSB |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16845934
|
Proteintech
16341-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16835934
|
Proteintech
16341-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a highly conserved enzyme that belongs to the aminoacyl-tRNA synthetase class IIc subfamily. This enzyme comprises the regulatory beta subunits that form a tetramer with two catalytic alpha subunits. In the presence of ATP, this tetramer is responsible for attaching L-phenylalanine to the terminal adenosine of the appropriate tRNA. A pseudogene located on chromosome 10 has been identified.Specifications
| FARSB | |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| Q9NSD9, Q9WUA2 | |
| 10056, 23874, 301544 | |
| FARSB Fusion Protein Ag9522 | |
| Primary | |
| -20°C | |
| FARSB |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| FARSB, FARSLB, FRSB, HSPC173, PheHB, PheRS | |
| Farsb | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title