Learn More
FARSA Rabbit anti-Human, Mouse, Rat, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against recombinant human FARSA.
Brand: Abnova PAB29320.100uL
Description
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. This gene encodes a product which is similar to the catalytic subunit of prokaryotic and Saccharomyces cerevisiae phenylalanyl-tRNA synthetases (PheRS). This gene product has been shown to be expressed in a tumor-selective and cell cycle stage- and differentiation-dependent manner, the first member of the tRNA synthetase gene family shown to exhibit this type of regulated expression [provided by RefSeq]
Sequence: EVIEAELRSTKHWELTAEGEEIAREGSHEARVFRSIPPEGLAQSELMRLPSGKVGFSKAMSNKWIRVDKSAADGPRVFRVVDSMEDEVQRRLQLVRGGQAEKLGEKERSELRKRKLLAEVTLKTYWVSKGSpecifications
| FARSA | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant human FARSA. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| FARSA | |
| Recombinant protein corresponding to amino acids of human FARSA. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:1000-1:2500) Western Blot (1:100-1:250) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| CML33/FARSL/FARSLA/FRSA/PheHA | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 2193 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.