missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FARSA Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA551657

Product Code. 15759610

  • £483.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: EVIEAELRST KHWELTAEGE EIAREGSHEA RVFRSIPPEG LAQSELMRLP SGKVGFSKAM SNKWIRVDKS AADGPRVFRV VDSMEDEVQR RLQLVRGGQA EKLGEKERSE LRKRKLLAEV TLKTYWVSKG Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.

Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. FARS2 is a phenylalanine-tRNA synthetase (PheRS) localized to the mitochondrion which consists of a single polypeptide chain, unlike the (alpha-beta)2 structure of the prokaryotic and eukaryotic cytoplasmic forms of PheRS. Structure analysis and catalytic properties indicate mitochondrial PheRSs may constitute a class of PheRS distinct from the enzymes found in prokaryotes and in the eukaryotic cytoplasm. Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. FARS1 encodes a mitochondrially-located phenylalanine-tRNA synthetase (PheRS) which consists of a single polypeptide chain unlike the (alpha-beta)2 structure of the prokaryotic and eukaryotic cytoplasmic forms of PheRS. Structure analysis and catalytic properties indicate mitochondrial PheRSs may constitute a class of PheRS distinct from the enzymes found in prokaryotes and the eukaryotic cytoplasm.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FARSA
Polyclonal
Unconjugated
FARSA
0610012A19Rik; CML33; FARS; Farsa; FARSL; FARSLA; FRSA; PheHA; phenylalanine tRNA ligase 1, alpha, cytoplasmic; phenylalanine--tRNA ligase alpha chain; phenylalanine--tRNA ligase alpha subunit; phenylalanine-tRNA synthetase alpha-subunit; phenylalanine-tRNA synthetase-like, alpha subunit; phenylalanyl-tRNA synthetase alpha chain; phenylalanyl-tRNA synthetase alpha subunit; phenylalanyl-tRNA synthetase, alpha subunit; phenylalanyl-tRNA synthetase-like, alpha subunit; pheRS
Rabbit
Antigen affinity chromatography
RUO
2193, 288917, 66590
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q505J8, Q8C0C7, Q9Y285
FARSA
Recombinant protein corresponding to Human FARSA. Recombinant protein control fragment (Product #RP-101902).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.