missing translation for 'onlineSavingsMsg'
Learn More

FARS2 Antibody, Novus Biologicals™

Product Code. 18390029 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390029 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18390029 Supplier Novus Biologicals Supplier No. H00010667D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FARS2 Polyclonal antibody specifically detects FARS2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen FARS2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_006558.1
Gene Alias dJ236A3.1, dJ520B18.2, dJ520B18.2 (FARS1 (phenylalanine-tRNA synthetase)), EC 6.1.1, EC 6.1.1.20, FARS1dJ236A3.1 (phenylalanine-tRNA synthetase), HSPC320, phenylalanine translase, phenylalanine tRNA ligase 2, mitochondrial, Phenylalanine--tRNA ligase, phenylalanine-tRNA synthetase 1 (mitochondrial), phenylalanine-tRNA synthetase 2 (mitochondrial), phenylalanyl-tRNA synthetase 2, mitochondrial, phenylalanyl-tRNA synthetase, mitochondrial, PheRS
Host Species Rabbit
Immunogen FARS2 (NP_006558.1, 1 a.a. - 451 a.a.) full-length human protein. MVGSALRRGAHAYVYLVSKASHISRGHQHQAWGSRPPAAECATQRAPGSVVELLGKSYPQDDHSNLTRKVLTRVGRNLHNQQHHPLWLIKERVKEHFYKQYVGRFGTPLFSVYDNLSPVVTTWQNFDSLLIPADHPSRKKGDNYYLNRTHMLRAHTSAHQWDLLHAGLDAFLVVGDVYRRDQIDSQHYPIFHQLEAVRLFSKHELFAGIKDGESLQLFEQSSRSAHKQETHTMEAVKLVEFDLKQTLTRLMAHLFGDELEIRWVDCYFPFTHPSFEMEINFHGEWLEVLGCGVMEQQLVNSAGAQDRIGWAFGLGLERLAMILYDIPDIRLFWCEDERFLKQFCVSNINQKVKFQPLSKYPAVINDISFWLPSENYAENDFYDLVRTIGGDLVEKVDLIDKFVHPKTHKTSHCYRITYRHMERTLSQREVRHIHQALQEAAVQLLGVEGRF
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 10667
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.