missing translation for 'onlineSavingsMsg'
Learn More

FARP1 Antibody, Novus Biologicals™

Product Code. 18376761 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18376761 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18376761 Supplier Novus Biologicals Supplier No. NBP276803

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

FARP1 Polyclonal specifically detects FARP1 in Human samples. It is validated for Flow Cytometry, Peptide ELISA, Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FARP1
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Polyclonal
Concentration 0.50 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 10 μ/mL, Peptide ELISA Detection Limit 1:32000, Immunofluorescence 10 μ/mL
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_005757, NP_001273768.1, NP_001001715.2
Gene Alias CDEPPleckstrin homology domain-containing family C member 2, Chondrocyte-derived ezrin-like protein, FERM, RhoGEF (ARHGEF) and pleckstrin domain protein 1 (chondrocyte-derived), FERM, RhoGEF and pleckstrin domain-containing protein 1, MGC87400, PLEKHC2PH domain-containing family C member 2
Gene Symbols FARP1
Host Species Goat
Immunogen Peptide with sequence GEIEQRPTPGSRL-C, from the N Terminus of the protein sequence according to NP_005757.1; NP_001001715.2; NP_001273768.1.
Quantity 0.1 mg
Primary or Secondary Primary
Gene ID (Entrez) 10160
Test Specificity This antibody is expected to recognize isoform 1 (NP_005757.1), isoform 2 (NP_001001715.2), and isoform 3 (NP_001273768.1)
Target Species Human
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.