missing translation for 'onlineSavingsMsg'
Learn More

FARP1 Antibody (2D4), Novus Biologicals™

Product Code. 18392649 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392649 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18392649 Supplier Novus Biologicals Supplier No. H00010160M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FARP1 Monoclonal antibody specifically detects FARP1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen FARP1
Applications Western Blot, ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2D4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005757
Gene Alias CDEPPleckstrin homology domain-containing family C member 2, Chondrocyte-derived ezrin-like protein, FERM, RhoGEF (ARHGEF) and pleckstrin domain protein 1 (chondrocyte-derived), FERM, RhoGEF and pleckstrin domain-containing protein 1, MGC87400, PLEKHC2PH domain-containing family C member 2
Host Species Mouse
Immunogen FARP1 (NP_005757.1, 471 a.a. ~ 549 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. TGSLTGSPHLSELSVNSQGGVAPANVTLSPNLSPDTKQASPLISPLLNDQACPRTDDEDEGRRKRFPTDKAYFIAKEVS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10160
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.