missing translation for 'onlineSavingsMsg'
Learn More

Faropenem Sodium Salt Hemipentahydrate, TRC

Product Code. 30390593
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390593 10 mg
30393091 25 mg
30389122 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390593 Supplier Toronto Research Chemicals Supplier No. TRCF10330010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 158365-51-6
Molecular Formula 2 C12 H14 N O5 S . 2 Na . 5 H2 O
Molecular Weight (g/mol) 704.67
Synonym 4-Thia-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid, 6-[(1R)-1-hydroxyethyl]-7-oxo-3-[(2R)-tetrahydro-2-furanyl]-, sodium salt, hydrate (2:2:5), (5R,6S)-, 4-Thia-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid, 6-(1-hydroxyethyl)-7-oxo-3-(tetrahydro-2-furanyl)-, monosodium salt, hydrate (2:5), [5R-[3(R*),5α,6α(R*)]]-, 4-Thia-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid, 6-[(1R)-1-hydroxyethyl]-7-oxo-3-[(2R)-tetrahydro-2-furanyl]-, monosodium salt, hydrate (2:5), (5R,6S)- (9CI), Faropenem sodium pentahemihydrate
IUPAC Name disodium;(5R,6S)-6-[(1R)-1-hydroxyethyl]-7-oxo-3-[(2R)-oxolan-2-yl]-4-thia-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate;pentahydrate
SMILES O.O.O.O.O.[Na+].[Na+].C[C@@H](O)[C@@H]1[C@H]2SC(=C(N2C1=O)C(=O)[O-])[C@H]3CCCO3.C[C@@H](O)[C@@H]4[C@H]5SC(=C(N5C4=O)C(=O)[O-])[C@H]6CCCO6

Specifications

Quantity 10 mg
InChI Formula InChI=1S/2C12H15NO5S.2Na.5H2O/c2*1-5(14)7-10(15)13-8(12(16)17)9(19-11(7)13)6-3-2-4-18-6;;;;;;;/h2*5-7,11,14H,2-4H2,1H3,(H,16,17);;;5*1H2/q;;2*+1;;;;;/p-2/t2*5-,6-,7+,11-;;;;;;;/m11......./s1
Formula Weight 704.1509
Chemical Name or Material Faropenem Sodium Salt, Hemipentahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.