missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Faropenem sodium, MedChemExpress
Description
Faropenem sodium is an orally bioavailable penem antibiotic which can efficiently kill Mycobacterium tuberculosis.
Specifications
Specifications
| Chemical Name or Material | Faropenem sodium |
| Physical Form | Solid |
| CAS | 122547-49-3 |
| Color | Yellow |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C12H14NNaO5S |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | H2O : 100 mg/mL (325.41 mM; Need ultrasonic) ∣DMSO : 25 mg/mL (81.35 mM; Need ultrasonic) |
| SMILES | O=C([C@@]([H])1[C@@H](C)O)N([C@@H]1S2)C(C(O[Na])=O)=C2[C@@H]3OCCC3 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?