missing translation for 'onlineSavingsMsg'
Learn More

Farnesyl Thiosalicylic Acid Amide (10 mg/mL in ethanol), TRC

Product Code. 30392810
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392810 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392810 Supplier Toronto Research Chemicals Supplier No. TRCF1025035ML
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Farnesyl Thiosalicylic Acid Amide
CAS 1092521-74-8
Molecular Formula C22H31NOS
Quantity 5 mL
Synonym 2-[[(2E,6E)-3,7,11-Trimethyl-2,6,10-dodecatrien-1-yl]thio]benzamide
InChI Formula InChI=1S/C22H31NOS/c1-17(2)9-7-10-18(3)11-8-12-19(4)15-16-25-21-14-6-5-13-20(21)22(23)24/h5-6,9,11,13-15H,7-8,10,12,16H2,1-4H3,(H2,23,24)/b18-11+,19-15+
SMILES CC(=CCC\C(=C\CC\C(=C\CSc1ccccc1C(=O)N)\C)\C)C
IUPAC Name 2-[(2E,6E)-3,7,11-trimethyldodeca-2,6,10-trienyl]sulfanylbenzamide
Molecular Weight (g/mol) 357.55
Formula Weight 357.2126
Concentration or Composition (by Analyte or Components) 10 mg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.