missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Farnesyl Pyrophosphate Triammonium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Farnesyl Pyrophosphate Triammonium Salt |
| CAS | 116057-57-9 |
| Molecular Formula | C15H37N3O7P2 |
| Quantity | 1 mg |
| Synonym | Diphosphoric Acid P-[(2E,6E)-3,7,11-Trimethyl-2,6,10-dodecatrien-1-yl] Ester Ammonium Salt, (E,E)-Diphosphoric Acid Mono(3,7,11-trimethyl-2,6,10-dodecatrienyl) Ester Triammonium Salt |
| InChI Formula | InChI=1S/C15H28O7P2.3H3N/c1-13(2)7-5-8-14(3)9-6-10-15(4)11-12-21-24(19,20)22-23(16,17)18;;;/h7,9,11H,5-6,8,10,12H2,1-4H3,(H,19,20)(H2,16,17,18);3*1H3/b14-9+,15-11+;;; |
| SMILES | N.N.N.CC(=CCC\C(=C\CC\C(=C\COP(=O)(O)OP(=O)(O)O)\C)\C)C |
| IUPAC Name | azane;phosphono [(2E,6E)-3,7,11-trimethyldodeca-2,6,10-trienyl] hydrogen phosphate |
| Molecular Weight (g/mol) | 433.42 |
| Formula Weight | 433.2107 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?