missing translation for 'onlineSavingsMsg'
Learn More

FAR2 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-97541PE

Product Code. 18638492

  • £309.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

FAR2 Polyclonal antibody specifically detects FAR2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FAR2
Polyclonal
Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
fatty acyl CoA reductase 2, FLJ10462, member 2, MLSTD1
Produced in rabbits immunized with E. coli-derived Human FAR2 fragment.
0.1 mL
Primary
Human
Purified
Immunocytochemistry, Immunohistochemistry (Paraffin)
PE
PBS
Rabbit
Antigen and protein A Affinity-purified
RUO
55711
Store at 4C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.