missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAPP2 Polyclonal Antibody

Brand: Thermo Scientific PA5113305
This item is not returnable.
View return policy
Description
Specifications
| FAPP2 | |
| Polyclonal | |
| Unconjugated | |
| PLEKHA8 | |
| 6330400G01; AA517676; BC052360; FAPP2; Fapp-2; hFAPP2; MGC3358; PH domain-containing family A member 8; phosphatidylinositol-four-phosphate adapter protein 2; phosphoinositol 4-phosphate adapter protein 2; pleckstrin homology domain containing A8; pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 8; pleckstrin homology domain-containing family A member 8; PLEKHA8; RGD1563940; Serologically defined breast cancer antigen NY-BR-86 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 231999, 84725 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q80W71, Q96JA3 | |
| PLEKHA8 | |
| Recombinant Human Pleckstrin homology domain-containing family A member 8 protein (1-300AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction