missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FAP ELISA Kit

Product Code. 15464983
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
1 set
Each
This item is not returnable. View return policy

Product Code. 15464983

Brand: Invitrogen™ EHFAP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

The Human FAP ELISA quantitates Hu FAP in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu FAP. Principle of the method The Hu FAP solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The protein encoded by this gene is a homodimeric integral membrane gelatinase belonging to the serine protease family. It is selectively expressed in reactive stromal fibroblasts of epithelial cancers, granulation tissue of healing wounds, and malignant cells of bone and soft tissue sarcomas. This protein is thought to be involved in the control of fibroblast growth or epithelial-mesenchymal interactions during development, tissue repair, and epithelial carcinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q12884
Assay Range 16.38 to 4000 pg/mL
Assay Sensitivity 12 pg/mL
Calibration Range 12 pg/mL to 4000 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Product Type ELISA
Protein Family angiogenesis; cell adhesion; dipeptidyl-peptidase activity; endopeptidase activity; endothelial cell migration; melanocyte apoptotic process; melanocyte proliferation; metalloendopeptidase activity; mitotic cell cycle arrest; negative regulation of cell proliferation involved in contact inhibition; negative regulation of extracellular matrix disassembly; negative regulation of extracellular matrix organization; peptidase activity; positive regulation of cell cycle arrest; positive regulation of execution phase of apoptosis; proteolysis; proteolysis involved in cellular protein catabolic process; regulation of collagen catabolic process; regulation of fibrinolysis; serine-type endopeptidase activity; serine-type peptidase activity
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA antibody pair detects human FAP. Other species not determined yet.
Sufficient For 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 2191
Gene Symbol FAP
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Angiogenesis
Sample Volume Plasma 1 μL, Serum 1 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Synonym DPPIVA, SIMP, FAP
Target FAP
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.