missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FANCM Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568191
This item is not returnable.
View return policy
Description
FANCM Polyclonal Antibody for Western Blot
The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group M. Alternative splicing results in multiple transcript variants.
Specifications
| FANCM | |
| Polyclonal | |
| Unconjugated | |
| FANCM | |
| AI427100; ATP-dependent RNA helicase FANCM; C730036B14Rik; D12Ertd364e; FAAP250; FANCM; Fanconi anemia complementation group M; Fanconi anemia group M protein; Fanconi anemia group M protein homolog; Fanconi anemia, complementation group M; fanconi anemia-associated polypeptide of 250 kDa; Kiaa1596; Protein FACM; Protein Hef ortholog; RGD1307897 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57697 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8IYD8 | |
| FANCM | |
| Synthesized peptide derived from human FANCM. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction