missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCM Antibody (CV5.1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50418CL1
This item is not returnable.
View return policy
Description
FANCM Monoclonal antibody specifically detects FANCM in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/Immunofluorescence
Specifications
| FANCM | |
| Monoclonal | |
| CoraFluor 1 | |
| ATP-dependent RNA helicase FANCM, EC 3.6.1, FAAP250EC 3.6.4.13, Fanconi anemia, complementation group M, Fanconi anemia-associated polypeptide of 250 kDa, KIAA1596Fanconi anemia group M protein, MGC176453, Protein FACM, Protein Hef ortholog | |
| His-tagged denatured FANCM (aa 1507-1679) made in E.coli | |
| 0.1 mL | |
| DNA Repair, Epigenetics | |
| 57697 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), KnockDown | |
| CV5.1 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction