missing translation for 'onlineSavingsMsg'
Learn More

FANCL Mouse anti-Human, Clone: 6E2B6, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66639-1-IG-150UL

 View more versions of this product

Product Code. 16838705

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group L. Alternative splicing results in two transcript variants encoding different isoforms.
Specifications

Specifications

FANCL
Monoclonal
2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9NW38
FANCL
FANCL Fusion Protein Ag6192
150 μL
Primary
Human
Antibody
IgG2a
Western Blot
6E2B6
Unconjugated
FANCL
FAAP43, FANCL, Fanconi anemia group L protein, PHF9, POG
Mouse
Protein A
RUO
55120
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.