missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38338-100ul
This item is not returnable.
View return policy
Description
FANCL Polyclonal antibody specifically detects FANCL in Mouse samples. It is validated for ELISA,Western Blot
Specifications
| FANCL | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA | |
| E3 ubiquitin-protein ligase FANCL, EC 6.3.2.-, FAAP43PHF9FLJ10335, Fanconi anemia group L protein, Fanconi anemia, complementation group L, Fanconi anemia-associated polypeptide of 43 kDa, PHD finger protein 9, Pog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-255 of human FANCL (NP_060532.2).,, Sequence:, MAVTEASLLRQCPLLLPQNRSKTVYEGFISAQGRDFHLRIVLPEDLQLKNARLLCSWQLRTILSGYHRIVQQRMQHSPDLMSFMMELKMLLEVALKNRQELYALPPPPQFYSSLIEEIGTLGWDKLVYADTCFSTIKLKAEDASGREHLITLKLKAKYPAESPDYFVDFPVPFCASWTPQSSLISIYSQFLAAIESLKAFWDVMDEIDEKTWVLEPEKPPRSATARRIALGNNVSINIEVDPRHPTMLPECFFLG | |
| 100 μL | |
| Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents | |
| 55120 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction