Learn More
FANCI Mouse anti-Human, Unconjugated, Clone: 1B10E6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67304-1-IG-20UL
This item is not returnable.
View return policy
Description
The FANCI antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FANCI. This antibody recognizes human antigen. The FANCI antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| FANCI | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot | |
| 1B10E6 | |
| Unconjugated | |
| 20 μL | |
| 55215 | |
| IgG1 |