missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCI Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-243AF647
This item is not returnable.
View return policy
Description
FANCI Polyclonal antibody specifically detects FANCI in Human samples. It is validated for Western Blot
Specifications
| FANCI | |
| Polyclonal | |
| Western Blot | |
| Fanconi anemia group I protein, Fanconi anemia, complementation group I, KIAA1794FLJ10719, Protein FACI | |
| A synthetic peptide mapping to a region between residues 200 and 250 of human KIAA1794/FLJ10719 using the numbering given in TrEMBL entry Q9NVI1 (GeneID 55215). | |
| 0.1 mL | |
| Neuroscience, Stem Cell Markers | |
| 55215 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction