missing translation for 'onlineSavingsMsg'
Learn More

FANCG Antibody (2C8), Novus Biologicals™

Product Code. 18360658 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18360658 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18360658 Supplier Novus Biologicals Supplier No. H00002189M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FANCG Monoclonal antibody specifically detects FANCG in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen FANCG
Applications Western Blot, ELISA, Immunocytochemistry, Immunoprecipitation, KnockDown
Classification Monoclonal
Clone 2C8
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:1000
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH11623
Gene Alias DNA repair protein XRCC9, FAG, Fanconi anemia group G protein, Fanconi anemia, complementation group G, Protein FACG, X-ray repair, complementing defective, in Chinese hamster, 9, XRCC9X-ray repair complementing defective repair in Chinese hamster cells 9
Host Species Mouse
Immunogen FANCG (AAH11623, 1 a.a. ~ 622 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSRQTTSVGSSCLDLWREKNDRLVRQAKVAQNSGLTLRRQQLAQDALEGLRGLLHSLQGLPAAVPVLPLELTVTCNFIILRASLAQGFTEDQAQDIQRSLERVLETQEQQGPRLEQGLRELWDSVLRASCLLPELLSALHRLVGLQAALWLSADRLGDLALLLETLNGSQSGASKDLLLLLKTWSPPAEELDAPLTLQDAQGLKDVLLTAFAYRQGLQELITGNPDKALSSLHEAASGLCPRPVLVQVYTALGSCHRKMGNPQRALLYLVAALKEGSAWGPPLLEASRLYQQLGDTTAELESLELLVEALNVPCSSKAPQFLIEVELLLPPPDLASPLHCGTQSQTKHILASRCLQTGRAGDAAEHYLDLLALLLDSSEPRFSPPPSPPGPCMPEVFLEAAVALIQAGRAQDALTLCEELLSRTSSLLPKMSRLWEDARKGTKELPYCPLWVSATHLLQGQAWVQLGAQKVAISEFSRCLELLFRATPEEKEQGAAFNCEQGCKSDAALQQLRAAALISRGLEWVASGQDTKALQDFLLSVQMCPGNRDTYFHLLQTLKRLDRRDEATALWWRLEAQTKGSHEDALWSLPLYLESYLSWIRPSDRDAFLEEFRTSLPKSCDL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents
Primary or Secondary Primary
Gene ID (Entrez) 2189
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.