missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCF Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-52057
This item is not returnable.
View return policy
Description
FANCF Polyclonal specifically detects FANCF in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| FANCF | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| FAF, Fanconi anemia group F protein, Fanconi anemia, complementation group F, MGC126856, Protein FACF | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2188 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.2 - 1 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| NP_073562.1 | |
| FANCF | |
| Peptide with sequence C-RPQEELEPGIHK corresponding to internal region according to NP_073562.1. | |
| 0.1 mg | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction