missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCE Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£154.00 - £434.00
Specifications
| Antigen | FANCE |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232901
|
Novus Biologicals
NBP3-38539-100ul |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30230264
|
Novus Biologicals
NBP3-38539-20ul |
20 μL |
£154.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FANCE Polyclonal antibody specifically detects FANCE in Mouse samples. It is validated for ELISA,Western BlotSpecifications
| FANCE | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents | |
| PBS (pH 7.3), 50% glycerol | |
| 2178 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| FACEFAE, Fanconi anemia group E protein, Fanconi anemia, complementation group E, Protein FACE | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-255 of human FANCE (NP_068741.1).,, Sequence:, MATPDAGLPGAEGVEPAPWAQLEAPARLLLQALQAGPEGARRGLGVLRALGSRGWEPFDWGRLLEALCREEPVVQGPDGRLELKPLLLRLPRICQRNLMSLLMAVRPSLPESGLLSVLQIAQQDLAPDPDAWLRALGELLRRDLGVGTSMEGASPLSERCQRQLQSLCRGLGLGGRRLKSPQAPDPEEEENRDSQQPGKRRKDSEEEAASPEGKRVPKRLRCWEEEEDHEKERPEHKSLESLADGGSASPIKDQP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title