missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCE Antibody (L5-P1E6*G5), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50400F
This item is not returnable.
View return policy
Description
FANCE Monoclonal specifically detects FANCE in Human samples. It is validated for ELISA, Immunohistochemistry.
Specifications
| FANCE | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| FANCE | |
| Ovalbumin-conjugated synthetic peptide - KAALKHLGP | |
| 0.1 mL | |
| Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents | |
| 2178 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| ELISA, Immunohistochemistry | |
| L5-P1E6*G5 | |
| ELISA, Immunohistochemistry | |
| FACEFAE, Fanconi anemia group E protein, Fanconi anemia, complementation group E, Protein FACE | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction