missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCE Antibody (L5-P1E6*G5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50400
This item is not returnable.
View return policy
Description
FANCE Monoclonal antibody specifically detects FANCE in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| FANCE | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| FACEFAE, Fanconi anemia group E protein, Fanconi anemia, complementation group E, Protein FACE | |
| Ovalbumin-conjugated synthetic peptide - KAALKHLGP | |
| 0.1 mL | |
| Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents | |
| 2178 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry | |
| L5-P1E6*G5 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction