missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FANCD2 Recombinant Rabbit Monoclonal Antibody (23GB5365), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | FANCD2 |
|---|---|
| Clone | 23GB5365 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088750
![]() |
Invitrogen™
MA552846 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089082
|
Invitrogen™
MA552847 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FANCD2 Recombinant Monoclonal antibody specifically detects FANCD2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| FANCD2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q6IV68, Q80V62, Q9BXW9 | |
| 211651, 2177, 312641 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5365 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2410150O07Rik; AU015151; BB137857; DKFZp762A223; FA4; FACD; FACD2; FAD; FAD2; FA-D2; FANCD; FANCD2; Fanconi anemia complementation group D2; Fanconi anemia D2 protein; Fanconi anemia group D2 protein; Fanconi anemia group D2 protein homolog; Fanconi anemia, complementation group D2; FLJ23826; Protein FACD2 | |
| A synthesized peptide derived from human FANCD2 (200-255AA). | |
| Primary | |
| -20°C | |
| FANCD2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title