Learn More
FANCD2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28619-1-AP-150UL
 View more versions of this product
Product Code. 16876195
  • £349.00 / 150µL
Description
The FANCD2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human FANCD2. This antibody recognizes human antigen. The FANCD2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
FANCD2
Polyclonal
Unconjugated
150 μL
2177
IgG
Western Blot, ELISA
400 μg/mL