missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCD2, p Ser222 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-502
This item is not returnable.
View return policy
Description
FANCD2 Polyclonal specifically detects FANCD2 in Human samples. It is validated for Western Blot.
Specifications
| FANCD2 (p Ser222) | |
| Polyclonal | |
| Western Blot 1:100-1:500 | |
| Q9BXW9 | |
| FANCD2 | |
| This FANCD2 [p Ser222] Antibody was developed against a synthetic peptide made to human FANCD2 (residues 217-226), surrounding the phosphorylated Serine 222. [Swiss-Prot# Q9BXW9] | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Antisera |
| Western Blot | |
| Unconjugated | |
| Whole antisera with 0.1% Sodium Azide | |
| DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3 | |
| Rabbit | |
| 164.1 kDa | |
| 0.2 mL | |
| Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents | |
| 2177 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction