missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCD2 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-182G
This item is not returnable.
View return policy
Description
FANCD2 Polyclonal specifically detects FANCD2 in Human, Mouse, Rat, Avian, Canine, Kangaroo, Primate, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting, Knockout Validated, Knockdown Validated.
Specifications
| FANCD2 | |
| Polyclonal | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting, Knockout Validated, Knockdown Validated | |
| Q9BXW9 | |
| FANCD2 | |
| This FANCD2 Antibody [DyLight 488] was developed against human FANCD2 fusion protein (N-terminal fragment). [Swiss-Prot #Q9BXW9] | |
| 0.05 mL | |
| Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents | |
| 2177 | |
| Human, Mouse, Rat, Avian, Canine, Kangaroo, Primate, Zebrafish | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot, Immunoassay, KnockDown | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is specific for FANCD2. | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction