missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCD2 Antibody (1290D) - BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
£151.00 - £332.00
Specifications
| Antigen | FANCD2 |
|---|---|
| Clone | 1290D |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 0.1 - 0.5 ug/ml, Simple Western 1 ug/ml, Flow Cytometry 1 ug/ml, Immunohistochemistry 5-10 ug/ml, Immunohistochemistry-Paraffin 5-10 ug/ml, Flow (Intracellular), Knockout Validated |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18268730
|
Novus Biologicals
NBP2-54808SS |
0.025 mg |
£151.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18207500
|
Novus Biologicals
NBP2-54808 |
0.1 mg |
£332.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FANCD2 Monoclonal specifically detects FANCD2 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular), Knockout Validated.Specifications
| FANCD2 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3 | |
| FANCD2 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| 164.1 kDa |
| 1290D | |
| Western Blot 0.1 - 0.5 ug/ml, Simple Western 1 ug/ml, Flow Cytometry 1 ug/ml, Immunohistochemistry 5-10 ug/ml, Immunohistochemistry-Paraffin 5-10 ug/ml, Flow (Intracellular), Knockout Validated | |
| Monoclonal | |
| Rabbit | |
| Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents | |
| PBS with 0.02% Sodium Azide | |
| 2177 | |
| This FANCD2 Antibody (1290D) was developed against human FANCD2 fusion protein (N-terminal fragment). [Swiss-Prot #Q9BXW9] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title