missing translation for 'onlineSavingsMsg'
Learn More

FANCD2 Antibody (1290C), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18407175
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407175 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407175 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP261930MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

FANCD2 Monoclonal antibody specifically detects FANCD2 in Human, Mouse, Rat samples. It is validated for Western Blot, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen FANCD2
Applications Western Blot, KnockDown
Classification Monoclonal
Clone 1290C
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Knockout Validated
Formulation 50mM Sodium Borate
Gene Alias DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3
Host Species Rabbit
Immunogen This FANCD2 Antibody (1290C) was developed against human FANCD2 fusion protein (N-terminal fragment). [Swiss-Prot #Q9BXW9]
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Genes Sensitive to DNA Damaging Agents
Primary or Secondary Primary
Gene ID (Entrez) 2177
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.