missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FANCA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105723
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FANCA.
This gene is a member of the synaptotagmin gene family and encodes a protein similar to other family members that are known calcium sensors and mediate calcium-dependent regulation of membrane trafficking in synaptic transmission. The encoded protein is also a substrate for ubiquitin-E3-ligase parkin. The gene has previously been referred to as synaptotagmin XII but has been renamed synaptotagmin XI to be consistent with mouse and rat official nomenclature.
Specifications
| FANCA | |
| Polyclonal | |
| Unconjugated | |
| FANCA | |
| AW208693; FA; FA1; FAA; FACA; FAH; FA-H; Fanca; FANCH; Fanconi anemia complementation group A; Fanconi anemia group A protein; Fanconi anemia group A protein homolog; Fanconi anemia, complementation group A; Fanconi anemia, complementation group H; Fanconi anemia, type 1; Protein FACA | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14087, 2175 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O15360, Q9JL70 | |
| FANCA | |
| A synthesized peptide derived from human FANCA(Accession O15360), corresponding to amino acid residues A1423-F1455. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction