Learn More
Invitrogen™ FANCA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596755
Description
Immunogen sequence: MSDSWVPNSA SGQDPGGRRR AWAELLAGRV KREKYNPERA QKLKESAVRL LRSHQDLNAL LLEVEGPLCK KLSLSKVIDC DSSEAYANHS SSFIGSALQD QASRLGVPVG ILSAGMVASS VGQICTAPAE TSHPVLLTVE QRKKLSSLLE FAQYLLAHSM FSRLSFCQEL WKIQSSLLLE AVWHLHVQGI VSLQELLESH PDMHAVGSWL FRNLCCLCEQ MEASCQHADV ARAMLSDFVQ MFVLRGFQKN SDLRRTVEPE KMPQVTVDVL QRMLI; Positive Samples: HeLa; Cellular Location: Cytoplasm, Nucleus.
This gene is a member of the synaptotagmin gene family and encodes a protein similar to other family members that are known calcium sensors and mediate calcium-dependent regulation of membrane trafficking in synaptic transmission. The encoded protein is also a substrate for ubiquitin-E3-ligase parkin. The gene has previously been referred to as synaptotagmin XII but has been renamed synaptotagmin XI to be consistent with mouse and rat official nomenclature.
Specifications
| FANCA | |
| Polyclonal | |
| Unconjugated | |
| FANCA | |
| AW208693; FA; FA1; FAA; FACA; FAH; FA-H; Fanca; FANCH; Fanconi anemia complementation group A; Fanconi anemia group A protein; Fanconi anemia group A protein homolog; Fanconi anemia, complementation group A; Fanconi anemia, complementation group H; Fanconi anemia, type 1; Protein FACA | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2175 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O15360 | |
| FANCA | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-275 of human FANCA (NP_0001262). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.