missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAN Polyclonal Antibody
GREENER_CHOICE

Product Code. 15702641 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15702641 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15702641 Supplier Invitrogen™ Supplier No. PA554707

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: FELDVPGKVE DVVETLLQLH RASCLDKLGD QTAMITAILQ SRLARTSFDK NRFQNISEKL HMECKAE Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.

This gene encodes a WD-repeat protein that binds the cytoplasmic sphingomyelinase activation domain of the 55kD tumor necrosis factor receptor. This protein is required for TNF-mediated activation of neutral sphingomyelinase and may play a role in regulating TNF-induced cellular responses such as inflammation. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NSMAF
Gene Accession No. Q92636
Gene Alias AA959567; C630007J05; factor associated with neutral sphingomyelinase activation; Factor associated with N-SMase activation; FAN; GRAMD5; neutral sphingomyelinase (N-SMase) activation associated factor; neutral sphingomyelinase activation associated factor; NSMAF; Protein FAN
Gene Symbols NSMAF
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAN. Recombinant protein control fragment (Product #RP-93731).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8439
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.