missing translation for 'onlineSavingsMsg'
Learn More

Famotidine, TRC

Product Code. 30405225
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 30405225

Brand: Toronto Research Chemicals TRCF102250500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Famotidine
CAS 76824-35-6
Purity Grade Notes HPLC
Molecular Formula C8 H15 N7 O2 S3
Quantity 500 mg
Synonym 3-[[[2-[(Aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]-N-(aminosulfonyl)-propanimidamide, 3-[(2-Diaminomethyleneaminothiazol-4-yl)methylthio]-N-sulfamoylpropionamidine, Agufam, Amfamox, Antodine, Apo-Famotidine, Apogastine, Bestidine, Brolin, Cepal, Dibrit 40, Digervin, Dispromil, Dispronil, Durater, Facid, Fadin, Fadine, Fadyn, Fagastine, Famo, FamoABZ, Famocid, Famodar, Famodil, Famodin, Famodine, Famogard, Famogast, Famonerton, Famopsin, Famos, Famosan, Famostagine-D, Famotal, Famotep, Famotidine, Famotin, Famotine, Famowal, Famox, Famoxal, Fanosin, Fanox, Farmotex, Fenox, Fibonel, Fudone, Fuweidin, Ganor, Ganor mite, Gaster, Gastridin, Gastrion, Gastro, Gastrodomina, Gastropen, H 2 Bloc, Ifada, Kemofam, Lecedil, Logos, MK 208, Mosul, Motiax, Muclox, N-(Aminosulfonyl)-3-[[[2-[(diaminomethylene)amino]-4-thiazolyl]methyl]thio]propanimidamide, Nevofam, Nulcerin, Panalba, Pepcid, Pepcid AC, Pepcid PM, Pepcidin, Pepcidin Rapitab, Pepcidina, Pepcidine, Pepdif, Pepdine, Pepdul, Pepdul mite, Pepfamin, Peptan, Peptifam, Pepzan, Purifam, Quamatel, Quamtel, Restadin, Rogasti, Sedanium R, Supertidine, Tamin, Topcid, Ulcatif, Ulcedine, Ulceran, Ulcetrax, Ulcidine, Ulcofam, Ulfagel, Ulfam, Ulfamid, Ulfinol, Ulped, Ulped AR, Weimok, YM 11170
InChI Formula InChI=1S/C8H15N7O2S3/c9-6(15-20(12,16)17)1-2-18-3-5-4-19-8(13-5)14-7(10)11/h4H,1-3H2,(H2,9,15)(H2,12,16,17)(H4,10,11,13,14)
SMILES NC(=Nc1nc(CSCCC(=NS(=O)(=O)N)N)cs1)N
IUPAC Name 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]-N'-sulfamoylpropanimidamide
Molecular Weight (g/mol) 337.45
Formula Weight 337.0449
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.