missing translation for 'onlineSavingsMsg'
Learn More

Famotidine Acetaldehyde Adduct Maleate (2-[4-[2-(3-Methyl-1,1-dioxo-3,4-dihydro-2H-1,2,4,6-thiatriazin-5-yl)ethylsulfanylmethyl]-1,3-thiazol-2-yl]guanidine Maleate), Mikromol™

Product Code. 17994456
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17994456 25 mg 25mg
17993756 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17994456 Supplier Mikromol Supplier No. MM0029.160025
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Famotidine Acetaldehyde Adduct Maleate (2-[4-[2-(3-Methyl-1,1-dioxo-3,4-dihydro-2H-1,2,4,6-thiatriazin-5-yl)ethylsulfanylmethyl]-1,3-thiazol-2-yl]guanidine Maleate)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Famotidine Acetaldehyde Adduct Maleate
CAS 2575516-65-1
Analyte or Component Names Famotidine Acetaldehyde Adduct Maleate
Shipping Condition Room Temperature
Molecular Formula C10 H17 N7 O2 S3 . C4 H4 O4
Certifications/Compliance ISO 9001
Synonym 2-(4-(((2-(3-Methyl-1,1-dioxido-3,4-dihydro-2H-1,2,4,6-thiatriazin-5-yl)ethyl)sulfanyl)methyl)-1,3-thiazol-2-yl)guanidine maleate, Famotidine Impurity C2 maleate, Famotidine acetaldehyde adduct maleate
InChI Formula InChI=1S/C10H17N7O2S3.C4H4O4/c1-6-13-8(17-22(18,19)16-6)2-3-20-4-7-5-21-10(14-7)15-9(11)12;5-3(6)1-2-4(7)8/h5-6,16H,2-4H2,1H3,(H,13,17)(H4,11,12,14,15);1-2H,(H,5,6)(H,7,8)/b;2-1-
SMILES CC1NC(=NS(=O)(=O)N1)CCSCc2csc(N=C(N)N)n2.OC(=O)\C=C/C(=O)O
IUPAC Name (Z)-but-2-enedioic acid;2-[4-[2-(3-methyl-1,1-dioxo-3,4-dihydro-2H-1,2,4,6-thiatriazin-5-yl)ethylsulfanylmethyl]-1,3-thiazol-2-yl]guanidine
Molecular Weight (g/mol) 479.55
Quantity 25 mg
Formula Weight 479.0715 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.