missing translation for 'onlineSavingsMsg'
Learn More

Famotidine, 98+%, Thermo Scientific™

A competitive histamine H2 receptor antagonist

£36.20 - £170.00

Chemical Identifiers

CAS 76824-35-6
Molecular Formula C8H15N7O2S3
Molecular Weight (g/mol) 337.435
MDL Number MFCD00079297
InChI Key XUFQPHANEAPEMJ-UHFFFAOYSA-N
Synonym famotidine, pepcid ac, apogastine, amfamox, antodine, bestidine, digervin, dispromil, famodil, gastridin
PubChem CID 5702160
ChEBI CHEBI:4975
IUPAC Name 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]-N'-sulfamoylpropanimidamide
SMILES C1=C(N=C(S1)N=C(N)N)CSCCC(=NS(=O)(=O)N)N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15415429
SDS View Documents
Thermo Scientific Alfa Aesar
J63165.03
1g
ONLINE EXCLUSIVE PRICE
£37.00
£36.20 / 1g
Save £0.80
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15425429
SDS View Documents
Thermo Scientific Alfa Aesar
J63165.06
5g
ONLINE EXCLUSIVE PRICE
£174.00
£170.00 / 5g
Save £4.00
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in water and methanol

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

76824-35-6
337.435
XUFQPHANEAPEMJ-UHFFFAOYSA-N
5702160
3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]-N'-sulfamoylpropanimidamide
C8H15N7O2S3
MFCD00079297
famotidine, pepcid ac, apogastine, amfamox, antodine, bestidine, digervin, dispromil, famodil, gastridin
CHEBI:4975
C1=C(N=C(S1)N=C(N)N)CSCCC(=NS(=O)(=O)N)N

Specifications

76824-35-6
MFCD00079297
famotidine, pepcid ac, apogastine, amfamox, antodine, bestidine, digervin, dispromil, famodil, gastridin
XUFQPHANEAPEMJ-UHFFFAOYSA-N
3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]-N'-sulfamoylpropanimidamide
5702160
337.45
Famotidine
C8H15N7O2S3
14,3930
Soluble in water and methanol
C1=C(N=C(S1)N=C(N)N)CSCCC(=NS(=O)(=O)N)N
337.435
CHEBI:4975
≥98%
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : UA2300000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.