missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris™ Famotidine

Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Famotidine is a potent and high affinity histamine H2 receptor biased inverse agonist (Kd = 14 nM, IC50 = 33 nM). Reduces basal receptor activity by 75%. Inhibits Gs-mediated pathways but acts as a partial agonist of β-arrestin recruitment (EC50 = 105 nM). Inhibits gastric acid secretion and histamine-induced mast cell activation. Also inhibits HIV replication in vitro.
Specifications
Specifications
| Chemical Name or Material | 3-[[[2-[(Aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]-N-(aminosulfonyl)propanimidamide |
| CAS | 76824-35-6 |
| Quantity | 100 mg |
| Target | Histamine H2 Receptor Antagonists |
| Molecular Formula | C8H15N7O2S3 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction